Learn More
Abcam SRT-3025 hydrochloride, 25MG
Catalog No. 502671218
Encompass_Preferred
MW 606.2 Da, Purity >=98%. SRT-3025 is a potent and selective Sirt1 activator. It inhibits RANKL-induced osteoclast differentiation, fusion and resorptive capacity without affecting osteoclast survival. SRT-3025 inhibits RANKL-induced osteoclastogenesis in bone marrow-derived macrophages (BMMs) by activating AMPK and deacetylating RelA/p65 lysine 310, critical for activation of the NF-κB signaling pathway.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1218 Supplier Abcam Supplier No. AB28702625MG
MW 606.2 Da, Purity >=98%. SRT-3025 is a potent and selective Sirt1 activator. It inhibits RANKL-induced osteoclast differentiation, fusion and resorptive capacity without affecting osteoclast survival. SRT-3025 inhibits RANKL-induced osteoclastogenesis in bone marrow-derived macrophages (BMMs) by activating AMPK and deacetylating RelA/p65 lysine 310, critical for activation of the NF-κB signaling pathway.
The product is subject to the following: Abcam Restricted Use Statement