Learn More
Cayman Chemical SRT 3025 (hydrochloride), Purity: ≥98%, CAS: 2070015-26-6, 100 mg
SDP
Catalog No. 502946469
Encompass_Preferred
A SIRT1 activator; increases SIRT1 levels in, and inhibits RANKL-induced osteoclastogenesis of, mouse BMDMs at 2 µM; inhibits proliferation of SU.86.86 cells (IC50 = 0.98 µM); decreases plasma levels of LDL, VLDL, and total cholesterol in ApoE-/- mice fed a high-cholesterol diet at 3.18 g/kg
Catalog No. 50-294-6469 Supplier Cayman Chemical Supplier No. 34420100 MG
A SIRT1 activator; increases SIRT1 levels in, and inhibits RANKL-induced osteoclastogenesis of, mouse BMDMs at 2 µM; inhibits proliferation of SU.86.86 cells (IC50 = 0.98 µM); decreases plasma levels of LDL, VLDL, and total cholesterol in ApoE-/- mice fed a high-cholesterol diet at 3.18 g/kg