Learn More
Selleck Chemical LLC SRT1720 HCl 50mg 1001645-58-4
SDP
Catalog No. 508141
Encompass_Preferred
SRT1720 HCl is a selective SIRT1 activator with EC50 of 0.16 ?M in a cell-free assay, but is >230-fold less potent for SIRT2 and SIRT3. SRT1720 induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-814-1 Supplier Selleck Chemical LLC Supplier No. S112950MG
SRT1720 HCl is a selective SIRT1 activator with EC50 of 0.16 ?M in a cell-free assay, but is >230-fold less potent for SIRT2 and SIRT3. SRT1720 induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.