Learn More
Apexbio Technology LLC SRT1720 HCl 1001645-58-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A41805.1
Encompass_Preferred
SRT1720 HCl is a small molecule activator of SIRT1 exerting more potent activation than resveratrol It functions through a SIRT1- and PGC-1 -dependent mechanism modulating mitochondrial oxidative metabolism and insulin sensitivity The compound has demonstrated activity in regulating cell growth and apoptosis in multiple myeloma (MM) cells without notably affecting normal cell viability Its antitumor properties relate to activation of caspase-8 caspase-9 caspase-3 and PARP enhancement of reactive oxygen species generation induction of ATM/CHK2 signaling suppression of NF- B signaling and reduction of VEGF-induced migration SRT1720 is utilized in biomedical research to investigate metabolic pathways tumor biology and potential sensitization effects to chemotherapy agents
Catalog No. 50-000-61450