Learn More
Abcam SRT1720 hydrochloride, 5MG
Catalog No. 502671873
Encompass_Preferred
MW 506 Da, Purity >=98%. Selective SIRT1 activator (EC₅₀ = 0.16 μM), 230-fold less potent for SIRT2 and SIRT3. Induce autophagy.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1873 Supplier Abcam Supplier No. AB2735985MG
MW 506 Da, Purity >=98%. Selective SIRT1 activator (EC₅₀ = 0.16 μM), 230-fold less potent for SIRT2 and SIRT3. Induce autophagy.
The product is subject to the following: Abcam Restricted Use Statement