Learn More
Abcam SRT1720 hydrochloride, 25MG
Catalog No. 502671874
Encompass_Preferred
MW 506 Da, Purity >=98%. Selective SIRT1 activator (EC₅₀ = 0.16 μM), 230-fold less potent for SIRT2 and SIRT3. Induce autophagy.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1874 Supplier Abcam Supplier No. AB27359825MG
MW 506 Da, Purity >=98%. Selective SIRT1 activator (EC₅₀ = 0.16 μM), 230-fold less potent for SIRT2 and SIRT3. Induce autophagy.
The product is subject to the following: Abcam Restricted Use Statement