Learn More
TARGETMOL CHEMICALS INC SRT1720 HYDROCHLORIDE 5MG
SDP
Catalog No. 502580831
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SRT1720 hydrochloride (SRT1720 HCl) is a selective activator of SIRT1 (EC1.5 0.16 uM) and shows less potent activities on SIRT2/SIRT3 (EC1.5s 37 uM/300 uM). purity: 99%
Catalog No. 50-258-0831 Supplier TARGETMOL CHEMICALS INC Supplier No. T24125MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SRT1720 hydrochloride (SRT1720 HCl) is a selective activator of SIRT1 (EC1.5 0.16 uM) and shows less potent activities on SIRT2/SIRT3 (EC1.5s 37 uM/300 uM). purity: 99%