Learn More
Sigma Aldrich Fine Chemicals Biosciences srum PlusTM II medum Sup500ML
Catalog No. 502905123
Encompass
Serum Plus(TM) II Medium Supplement contains a low level of fetal Bovine Serum (fBS) in a proprietary formulation to promote cell attachment and cell growth.
Catalog No. 50-290-5123 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 14009C500ML
Add to Cart
Add to Cart
Serum Plus(TM) II Medium Supplement contains a low level of fetal Bovine Serum (fBS) in a proprietary formulation to promote cell attachment and cell growth.