Learn More
Medchemexpress LLC HY-105685 5mg Medchemexpress, SRX246 CAS:512784-93-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1056855MG
Encompass_Preferred
Medchemexpress, HY-105685 5mg SRX246 CAS:512784-93-9 SRX246 is a potent, CNS-penetrant, highly selective, orally bioavailable vasopressin 1a (V1a) receptor antagonist (Ki=0.3 nM for human V1a). SRX246 has no interaction at V1b and V2 receptors. SRX246 also displays negligible binding at 64 others receptors classes, including 35 G-proteincoupled receptors. SRX246 can be used for treatment of stress-related disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8288