Learn More
Selleck Chemical LLC SRX3177 E4702-1G
SDP
Catalog No. 5000041977
Encompass_Preferred
SRX3177 is a potent triple inhibitor targeting CDK4/6 PI3K and BRD4 with IC50 values of 33 nM for BRD4 BD1 89 nM for BRD4 BD2 79 nM for PI3K 83 nM for PI3K 3 18 M for PI3K 2 5 nM for CDK4 and 3 3 nM for CDK6 By simultaneously inhibiting these key pathways SRX3177 disrupts cancer cell signalling and exhibits significant cytotoxic effects in tumors
Catalog No. 50-000-41977 Supplier Selleck Chemical LLC Supplier No. E47021G
SRX3177 is a potent triple inhibitor targeting CDK4/6 PI3K and BRD4 with IC50 values of 33 nM for BRD4 BD1 89 nM for BRD4 BD2 79 nM for PI3K 83 nM for PI3K 3 18 M for PI3K 2 5 nM for CDK4 and 3 3 nM for CDK6 By simultaneously inhibiting these key pathways SRX3177 disrupts cancer cell signalling and exhibits significant cytotoxic effects in tumors