Learn More
Aviva Systems Biology Corp SRY Antibody - middle region (ARP34794_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34794_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRY. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SRY. Alternate names: TDF, TDY, SRXX1, SRXY1ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-55565