Learn More
Phoenix Metal Products Inc SS-100 MTL SLIDE STORAGE CAB
SDP
Catalog No. NC0322038
Encompass
Holds up to 5,000 1 in. x 3 in. (25mm x 75mm) microscope slides. Durable steel construction stacks up to 15 cabinets high for up to 75,000 slides per stack. 6 removable drawers per cabinet, with cardholders and a full length slide row separator.
Catalog No. NC0322038 Supplier Phoenix Metal Products Inc Supplier No. SS100
Holds up to 5,000 1 in. x 3 in. (25mm x 75mm) microscope slides. Durable steel construction stacks up to 15 cabinets high for up to 75,000 slides per stack. 6 removable drawers per cabinet, with cardholders and a full length slide row separator.