Learn More
HISCO SS 18 GAUGE NDL W/ PINK LOCK
SDP
Catalog No. NC0475010
Encompass
This item has a minimum qty of 5 per supplier requirements.
Stainless Steel 18 Gauge Needle with Pink Luer Lock Hub, .038" I.D, .050" O.D, 1" Long, 50/PK. The Weller KDS181P-12200 is a straight, 18 gauge, 1" long, pink dispensing needle. The needle features a stainless steel dispensing tip and plastic hub and is designed to be used with Luer Lok style syringes on a variety of general purpose fluid dispensing applications.
Catalog No. NC0475010 Supplier HISCO Supplier No. KDS181P12200
This item has a minimum qty of 5 per supplier requirements.
Stainless Steel 18 Gauge Needle with Pink Luer Lock Hub, .038" I.D, .050" O.D, 1" Long, 50/PK. The Weller KDS181P-12200 is a straight, 18 gauge, 1" long, pink dispensing needle. The needle features a stainless steel dispensing tip and plastic hub and is designed to be used with Luer Lok style syringes on a variety of general purpose fluid dispensing applications.