Learn More
New Jersey Fluid Systems LLC Swagelok SS-43GS4 Ball Valve, 316 Stainless Steel, 3000 psig Max Pressure, 1/4 in
Catalog No. NC1437438
Encompass
The Swagelok SS-43GS4 is a 1-piece ball valve designed for high performance in industrial and scientific applications. Made from 316 stainless steel, it offers a maximum pressure of 3000 psig and a temperature range of -65 to 300°F. Its encapsulated PTFE packing ensures reliable operation and minimizes adjustments, making it ideal for controlling flow with precision. The innovative design supports bi-directional flow, enhancing efficiency in system performance.
Catalog No. NC1437438 Supplier New Jersey Fluid Systems LLC Supplier No. SS43GS4
The Swagelok SS-43GS4 is a 1-piece ball valve designed for high performance in industrial and scientific applications. Made from 316 stainless steel, it offers a maximum pressure of 3000 psig and a temperature range of -65 to 300°F. Its encapsulated PTFE packing ensures reliable operation and minimizes adjustments, making it ideal for controlling flow with precision. The innovative design supports bi-directional flow, enhancing efficiency in system performance.