Learn More
Analytical Sales & Services SS 2 Position, 6 Port Prep Syringe Load Prep Manual Sample Injector with Built In Position Sensing Switch
Catalog No. 5000804396
Encompass
Stainless Steel 2 Position, 6 Port Prep Syringe Load Prep Manual Sample Injector with Built In Position Sensing Switch and Attached 10mL Sample Loop.\nInject from 10 - 100mL/min. Set for 4,000psi. Adjustable to 5,000psi.\n\nQty/Pkg: EACH
Catalog No. 50-008-04396 Supplier Analytical Sales & Services Supplier No. 43725I0384
Stainless Steel 2 Position, 6 Port Prep Syringe Load Prep Manual Sample Injector with Built In Position Sensing Switch and Attached 10mL Sample Loop.\nInject from 10 - 100mL/min. Set for 4,000psi. Adjustable to 5,000psi.\n\nQty/Pkg: EACH