Learn More
TARGETMOL CHEMICALS INC SS-208 5MG
SDP
Catalog No. 502581137
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. SS-208 is a selective inhibitor of HDAC6 (IC50 = 12 nM) with anti-tumor activity. SS-208 shows selectivity over other HDAC subtypes. purity: 99%
Catalog No. 50-258-1137 Supplier TARGETMOL CHEMICALS INC Supplier No. T169365MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. SS-208 is a selective inhibitor of HDAC6 (IC50 = 12 nM) with anti-tumor activity. SS-208 shows selectivity over other HDAC subtypes. purity: 99%