Learn More
New Jersey Fluid Systems LLC SS-2C-1/3, 1/8IN OD TUBE FIT
Supplier:  New Jersey Fluid Systems LLC SS2C1/3
Encompass
SS-2C-1/3, Stainless Steel "C" Series Check Valve, 1/8in OD Tube Fitting Ends, 1/3PSI Spring, FKM Seal
Catalog No. NC1949449