Learn More
AVANTOR MASTERFLEX (VWR LLC) Parker Hannifin 4F-MB6XPFA-SSP three-way ball valve, stainless steel, 1/4"" NPT(F)
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3052760
Encompass
Ball valves are designed primarily for on/off service. The three-way valve model has three ports, and a combination of the ports can be either open or closed for various flow pattern capabilities. The handle of the valve can be turned to change the flow pattern or on/off service of the valve. Choose from stainless steel/PFA or brass/PFA valve options.
Catalog No. NC0489283