Learn More
New Jersey Fluid Systems LLC SS-400-R-8, 1/4IN TUBE OD
Catalog No. NC1963880
Encompass
Stainless Steel Swagelok Tube Fitting, Reducer, 1/4 in. Tube OD x 1/2 in. Swagelok Tube Adapter
Catalog No. NC1963880 Supplier New Jersey Fluid Systems LLC Supplier No. SS400R8
Stainless Steel Swagelok Tube Fitting, Reducer, 1/4 in. Tube OD x 1/2 in. Swagelok Tube Adapter