Learn More
Ballymore Stainless Steel Fold and Store Ladder, 6 Step, 30" Wide Base, 14" Deep Top Step, Perforated Tread
Catalog No. NC3404884
Encompass_Preferred
Stainless Steel Rolling Ladder450 Lb. CapacityIdeal For Wash Down And Corrosive EnvironmentsType T304 Stainless Steel Construction4-12 Step Ladders Have 14" Deep Top Step StandardBead Blasted For Uniform FinishSpring Loaded Casters Are Stainless And Springs Are Carbon SteelComplies With Applicable OSHA & ANSI StandardsAll Stainless Ladders Are Manufactured With Industrial Grade Welds, Not Recommended For Clean Rooms. Ships Unassembled In A Box
Catalog No. NC3404884 Supplier Ballymore Supplier No. SSFAWL6P
Stainless Steel Rolling Ladder450 Lb. CapacityIdeal For Wash Down And Corrosive EnvironmentsType T304 Stainless Steel Construction4-12 Step Ladders Have 14" Deep Top Step StandardBead Blasted For Uniform FinishSpring Loaded Casters Are Stainless And Springs Are Carbon SteelComplies With Applicable OSHA & ANSI StandardsAll Stainless Ladders Are Manufactured With Industrial Grade Welds, Not Recommended For Clean Rooms. Ships Unassembled In A Box