Learn More
Analytical Sales & Services SS Adapter, 1/4-28 Male to 10-32 Female
Catalog No. 5000804480
Encompass
SS Adapter .040" ID 10-32 Female to 1/4-28 Male\n\nQty/Pkg: EACH
Catalog No. 50-008-04480 Supplier Analytical Sales & Services Supplier No. 415914
SS Adapter .040" ID 10-32 Female to 1/4-28 Male\n\nQty/Pkg: EACH