Learn More
Chemglass Life Sciences Stainless steel adapter, M30 female to 3/4 in male NPT (for circulator fittings)
SDP
Catalog No. NC2306853
Encompass
Stainless steel adapter that converts an M30 female port to a 3/4 in male NPT thread, designed for circulator fittings and laboratory fluid-transfer connections. Allows adaptation of M30-style circulator ports to standard 3/4 in NPT plumbing while maintaining a corrosion-resistant connection.
  • Converts M30 female to 3/4 in male NPT.
  • Made from stainless steel for corrosion resistance.
  • Enables secure, leak-resistant connections between circulator ports and NPT fittings.
  • Compact design for easy installation and tightening.
  • Suited for laboratory circulators and fluid-transfer systems.
Catalog No. NC2306853 Supplier Chemglass Life Sciences Supplier No. CG1969M108
Stainless steel adapter that converts an M30 female port to a 3/4 in male NPT thread, designed for circulator fittings and laboratory fluid-transfer connections. Allows adaptation of M30-style circulator ports to standard 3/4 in NPT plumbing while maintaining a corrosion-resistant connection.
  • Converts M30 female to 3/4 in male NPT.
  • Made from stainless steel for corrosion resistance.
  • Enables secure, leak-resistant connections between circulator ports and NPT fittings.
  • Compact design for easy installation and tightening.
  • Suited for laboratory circulators and fluid-transfer systems.