Learn More
Analytical Sales & Services SS Adjustable Flow Splitter with -01 Resistor, Post Column, Ratio 50:1 To 1000:1
Catalog No. NC1367837
Encompass
with -01 resistor, Post Column, Ratio 50:1 to 1000:1 (62-411)
Catalog No. NC1367837 Supplier Analytical Sales & Services Supplier No. 62411
with -01 resistor, Post Column, Ratio 50:1 to 1000:1 (62-411)