Learn More
MSC Miller ShadowLite Adjustable Beam Anchor, Stainless Steel and Aluminum, 4 in. to 12 in. Flange, 1 ea
Catalog No. 5000508276
Encompass
The Shadow series adjustable beam anchor is a durable, reusable fall protection device designed for use on I- and W-beams. Engineered for safety and mobility, this anchor is designed to slide along the beam as the worker moves. It is constructed from high-quality stainless steel, aluminum, and drop forged steel, and it meets all applicable ANSI and OSHA standards.
  • Fits flange widths from 4 in. (100 mm) to 12 in. (305 mm)
  • Accommodates flange thicknesses up to 1-1/4 in.
  • Suitable for use on top or bottom beam flanges
  • Features hand-retractable plunger for easy adjustment
  • Hardware meets ASTM 50-hour salt spray test requirements
  • Maximum working load of 310 lbs. (140.62 kg)
  • Minimum tensile strength of 5,000 lbs. (22 kN)
Catalog No. 50-005-08276 Supplier MSC Supplier No. 52873262
The Shadow series adjustable beam anchor is a durable, reusable fall protection device designed for use on I- and W-beams. Engineered for safety and mobility, this anchor is designed to slide along the beam as the worker moves. It is constructed from high-quality stainless steel, aluminum, and drop forged steel, and it meets all applicable ANSI and OSHA standards.
  • Fits flange widths from 4 in. (100 mm) to 12 in. (305 mm)
  • Accommodates flange thicknesses up to 1-1/4 in.
  • Suitable for use on top or bottom beam flanges
  • Features hand-retractable plunger for easy adjustment
  • Hardware meets ASTM 50-hour salt spray test requirements
  • Maximum working load of 310 lbs. (140.62 kg)
  • Minimum tensile strength of 5,000 lbs. (22 kN)