Learn More
MSC Honeywell Miller Shadow Beam Anchor, Stainless Steel, 12 to 24 in. Flange, 1 ea
Catalog No. 5000508950
Encompass
This fixed beam anchor is designed for fall protection applications, providing a secure connection point on structural beams. It is constructed from durable stainless steel and aluminum materials, featuring a hand-retractable plunger and a robust D-ring attachment for safety and reliability.
  • Fits beam flange widths from 10.2 cm to 61 cm (4 in. to 24 in.)
  • Accommodates beam flange thickness up to 6.4 cm (2.5 in.)
  • Features stainless steel support shaft and retractable plunger
  • D-ring attachment constructed of plated, drop-forged steel and stainless steel
  • Hardware meets ASTM fifty-hour salt spray test requirements
  • Certified to ANSI A10.14 and Z359.1 standards
  • Minimum tensile strength of 5,000 lb.
Catalog No. 50-005-08950 Supplier MSC Supplier No. 46505830
This fixed beam anchor is designed for fall protection applications, providing a secure connection point on structural beams. It is constructed from durable stainless steel and aluminum materials, featuring a hand-retractable plunger and a robust D-ring attachment for safety and reliability.
  • Fits beam flange widths from 10.2 cm to 61 cm (4 in. to 24 in.)
  • Accommodates beam flange thickness up to 6.4 cm (2.5 in.)
  • Features stainless steel support shaft and retractable plunger
  • D-ring attachment constructed of plated, drop-forged steel and stainless steel
  • Hardware meets ASTM fifty-hour salt spray test requirements
  • Certified to ANSI A10.14 and Z359.1 standards
  • Minimum tensile strength of 5,000 lb.