Learn More
Analytical Sales & Services SS Analytical Flow 350µL Static Bed Mixer - Cartridge Only
Catalog No. 5000806959
Encompass
Advantage Stainless Steel Analytical Flow 350µL Static Bed Mixer Cartridge Only\n\nQty/Pkg: EACH
Catalog No. 50-008-06959 Supplier Analytical Sales & Services Supplier No. 130003501
Advantage Stainless Steel Analytical Flow 350µL Static Bed Mixer Cartridge Only\n\nQty/Pkg: EACH