Learn More
Grainger SS Band Heavy Duty Steel 1-1/4
Catalog No. 502768286
Encompass_Preferred
The Grainger 16P351 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-woven fibers in a medium grade for cleaning and scouring. This 6 inch by 9 inch hand pad has an open lapping that exposes the fibers for scrubbing and is ideal for cleaning pots, pans, dishes, floors, walls, grills and more. It effectively removes grease, grime, food residue and more from a variety of surfaces. The Scotch-Brite pad is durable, lint-free and can be used wet or dry without scratching.
Catalog No. 50-276-8286 Supplier Grainger Supplier No. 16P351
The Grainger 16P351 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-woven fibers in a medium grade for cleaning and scouring. This 6 inch by 9 inch hand pad has an open lapping that exposes the fibers for scrubbing and is ideal for cleaning pots, pans, dishes, floors, walls, grills and more. It effectively removes grease, grime, food residue and more from a variety of surfaces. The Scotch-Brite pad is durable, lint-free and can be used wet or dry without scratching.