Learn More
Grainger SS Band Heavy Duty Steel 3/4
Catalog No. 502768632
Encompass_Preferred
The Band-It 0.044" Wide x 0.015" Thick 201 Stainless Steel Band is a thin stainless steel band that can be used for bundling, banding, securing, and fastening. It is constructed from 201 stainless steel for strength and corrosion resistance. The band has a width of 0.044" and thickness of 0.015". It can be tensioned and cinched around items using a separate tensioning tool. The stainless steel composition makes this band durable for indoor and outdoor use. With its narrow width, it can bundle small items or wires neatly. This is an economical stainless steel band option for general purpose bundling and securing tasks.
Catalog No. 50-276-8632 Supplier Grainger Supplier No. 16P349
The Band-It 0.044" Wide x 0.015" Thick 201 Stainless Steel Band is a thin stainless steel band that can be used for bundling, banding, securing, and fastening. It is constructed from 201 stainless steel for strength and corrosion resistance. The band has a width of 0.044" and thickness of 0.015". It can be tensioned and cinched around items using a separate tensioning tool. The stainless steel composition makes this band durable for indoor and outdoor use. With its narrow width, it can bundle small items or wires neatly. This is an economical stainless steel band option for general purpose bundling and securing tasks.