Learn More
Grainger SS Band Standard Duty Steel 5/8
Catalog No. 502765538
Encompass_Preferred
The SS Band Standard Duty Steel 5/8" is a steel band made from stainless steel for securing objects. It has a width of 5/8", a thickness of 0.020", and a tensile strength of 120,000 psi. The band has a smooth finish and comes in coils of 25 ft, 50 ft, and 100 ft lengths. It meets military, federal, and ASTM specifications for quality. The band can withstand temperatures up to 1000°F. It is commonly used for bundling, securing pipes, and general purpose holding.
Catalog No. 50-276-5538 Supplier Grainger Supplier No. 16P346
The SS Band Standard Duty Steel 5/8" is a steel band made from stainless steel for securing objects. It has a width of 5/8", a thickness of 0.020", and a tensile strength of 120,000 psi. The band has a smooth finish and comes in coils of 25 ft, 50 ft, and 100 ft lengths. It meets military, federal, and ASTM specifications for quality. The band can withstand temperatures up to 1000°F. It is commonly used for bundling, securing pipes, and general purpose holding.