Learn More
Cole-Parmer Cole-Parmer Stainless-steel Griffin-style beaker with easy-pour rim, 2000 mL
Catalog No. NC1745876 Shop All Cole-Parmer Products
Encompass_Preferred
These Griffin-style beakers with pouring spout have full capacity mark near the top to assure accurate measuring. Constructed of medium gauge 304 stainless steel, these beakers are durable and easy to clean.
Catalog No. NC1745876 Supplier Cole-Parmer Supplier No. 0720570
Add to Cart
Add to Cart
These Griffin-style beakers with pouring spout have full capacity mark near the top to assure accurate measuring. Constructed of medium gauge 304 stainless steel, these beakers are durable and easy to clean.