Learn More
Electron Microscopy Sciences EMS Vibrating Microtome Stainless Steel Blades
SDP
Catalog No. 50949617
Encompass_Preferred
Blades for EMS7000smz and EMS5000mz Vibrating Microtome are double beveled on both faces, honed to an acute cutting edge and hardened to 56 Rockwell. Relatively soft and usually used once or at a maximum changed every day.
Catalog No. 50-949-617 Supplier Electron Microscopy Sciences Supplier No. 75501SS
Blades for EMS7000smz and EMS5000mz Vibrating Microtome are double beveled on both faces, honed to an acute cutting edge and hardened to 56 Rockwell. Relatively soft and usually used once or at a maximum changed every day.