Learn More
Imtakt SS-C18 SCHERZO 4.6X150MM 3UM
Catalog No. NC0193659
Encompass
The Scherzo family of columns is one of the most unique and versatile series of columns on the market today. These multi-mode columns utilize reversed phase, cation and anion exchange, as well as normal phase modes all on the same column, virtually eliminating the need for ion-pairing reagents. -Polar and non-polar retention in Reversed Phase conditions Multi-level retention without ion-pairing reagents- Compatible with LC-MS- Most unique selectivity of any HPLC column on the market today- Wide range of method development possibilities
Catalog No. NC0193659 Supplier Imtakt Supplier No. SS005
The Scherzo family of columns is one of the most unique and versatile series of columns on the market today. These multi-mode columns utilize reversed phase, cation and anion exchange, as well as normal phase modes all on the same column, virtually eliminating the need for ion-pairing reagents. -Polar and non-polar retention in Reversed Phase conditions Multi-level retention without ion-pairing reagents- Compatible with LC-MS- Most unique selectivity of any HPLC column on the market today- Wide range of method development possibilities