Learn More
Uline Stainless Steel Cart, 2 shelf, 42 x 24 x 35""""
Catalog No. NC1930547
Encompass
All-welded, corrosion-resistant stainless steel.Lipped shelves, 1 1/2" deep.Shelves are 25" apart for supplies.5" polyurethane casters: 2 swivel, 2 rigid.
Catalog No. NC1930547 Supplier Uline Supplier No. H3352
All-welded, corrosion-resistant stainless steel.Lipped shelves, 1 1/2" deep.Shelves are 25" apart for supplies.5" polyurethane casters: 2 swivel, 2 rigid.