Learn More
McMaster-Carr SS CLAMP W WING NUT 1-1.5"
Catalog No. NC1672037
Encompass
High-Polish Quick-Clamp Sanitary Tube Fitting, 304 Stainless Steel Clamp with Wing Nut for 1" and 1-1/2" Tube OD
Catalog No. NC1672037 Supplier McMaster-Carr Supplier No. 4322K152
High-Polish Quick-Clamp Sanitary Tube Fitting, 304 Stainless Steel Clamp with Wing Nut for 1" and 1-1/2" Tube OD