Learn More
McMaster-Carr SS CLAMP W WING NUT 1/2-3/4"
Supplier:  McMaster-Carr 45195K44
Encompass
High-Polish Quick-Clamp Sanitary Tube Fitting, 304 Stainless Steel Clamp with Wing Nut, for 1/2 and 3/4" Tube OD
Catalog No. NC1672034