Learn More
MSC PFERD Small Cleaning Brush, 2x9 Rows, .006 Stainless Steel Wire, Wooden Block
Catalog No. 502839481
Encompass
This cleaning brush is designed for a wide range of applications, including cleaning spark plugs, small workpieces, and various tools. Constructed with 302 stainless steel wire, it offers excellent corrosion resistance and optimal brush life, making it particularly effective for use on stainless steel (INOX) materials.
  • Filament material: Stainless steel wire (INOX)
  • Filament diameter: .006 in
  • Brush configuration: 2 rows by 9 rows
  • Overall length: 8-5/8 in
  • Trim length: 1/2 in
  • Width: 2-1/2 in
  • Block width: 1/2 in
  • Manual application
  • Ideal for cleaning small weld seams and fillet welds
  • Suitable for aluminum, bronze, cobalt-based alloys, other non-ferrous metals, and stainless steel (INOX)
  • Applications: Cleaning, derusting, paint stripping, removing heat discolouration, removing oxide layers, roughening, and work on weld seams
Catalog No. 50-283-9481 Supplier MSC Supplier No. 66303348
This cleaning brush is designed for a wide range of applications, including cleaning spark plugs, small workpieces, and various tools. Constructed with 302 stainless steel wire, it offers excellent corrosion resistance and optimal brush life, making it particularly effective for use on stainless steel (INOX) materials.
  • Filament material: Stainless steel wire (INOX)
  • Filament diameter: .006 in
  • Brush configuration: 2 rows by 9 rows
  • Overall length: 8-5/8 in
  • Trim length: 1/2 in
  • Width: 2-1/2 in
  • Block width: 1/2 in
  • Manual application
  • Ideal for cleaning small weld seams and fillet welds
  • Suitable for aluminum, bronze, cobalt-based alloys, other non-ferrous metals, and stainless steel (INOX)
  • Applications: Cleaning, derusting, paint stripping, removing heat discolouration, removing oxide layers, roughening, and work on weld seams