Learn More
Eagle Stainless Container Eagle Stainless, 60 Liter Container with Lid and Gasket, 316L Stainless Steel. (CTH Series)
SDP
Catalog No. NC1293032
Encompass_Preferred
60 Liter container with clip down lid and platinum cured silicone gasket. Type 316L stainless steel. Pharmaceutical grade finish of 10 Ra or better.
Catalog No. NC1293032 Supplier Eagle Stainless Container Supplier No. CTH43316L
60 Liter container with clip down lid and platinum cured silicone gasket. Type 316L stainless steel. Pharmaceutical grade finish of 10 Ra or better.