Learn More
MSC Sheila Shine SS Cleaner, Liquid Aerosol, Wintergreen, 10 oz
Catalog No. 502866922
Encompass
This stainless steel cleaner and polish is formulated to remove various residues and restore shine to stainless steel surfaces. Its mild acetone-based formula effectively tackles dust, stains, rust, grease, water marks, and grime.
  • Formulated as a liquid aerosol
  • Mild acetone-based formula
  • Effectively removes dust, stains, rust, grease, water marks, and grime
  • Features a wintergreen scent
  • Supplied in a 10 fl oz aerosol can
Catalog No. 50-286-6922 Supplier MSC Supplier No. 14019145
This stainless steel cleaner and polish is formulated to remove various residues and restore shine to stainless steel surfaces. Its mild acetone-based formula effectively tackles dust, stains, rust, grease, water marks, and grime.
  • Formulated as a liquid aerosol
  • Mild acetone-based formula
  • Effectively removes dust, stains, rust, grease, water marks, and grime
  • Features a wintergreen scent
  • Supplied in a 10 fl oz aerosol can