Learn More
Strem, An Ascensus Company CAS 405235-55-4. 500mg. ((4S5S)-(-)-O-[1-Benzyl-1-(5-methyl-2-phenyl-45-dihydrooxazol-4-yl)-2-phenylethyl]-diphenylphosphinite)(15-COD)iridium(I) tetrakis(35-bis(trifluoromethyl)phenylborate min. 97% (SS)-[COD]Ir[Ph2PThrePHOX]. MFCD06658140
SDP
Catalog No. 502110857
Encompass
CAS 405235-55-4. 500mg. ((4S5S)-(-)-O-[1-Benzyl-1-(5-methyl-2-phenyl-45-dihydrooxazol-4-yl)-2-phenylethyl]-diphenylphosphinite)(15-COD)iridium(I) tetrakis(35-bis(trifluoromethyl)phenylborate min. 97% (SS)-[COD]Ir[Ph2PThrePHOX]. MFCD06658140. Molecular Weight 1719.25. Molecular Formula Ir(C8H12)(C37H34NO2P)B[C6H3(CF3)2]4ae. Color/form orange pwdr. Strem 77-5020. http//www.strem.com/catalog/v/77-5020/
Catalog No. 50-211-0857 Supplier Strem, An Ascensus Company Supplier No. 775020500MG
CAS 405235-55-4. 500mg. ((4S5S)-(-)-O-[1-Benzyl-1-(5-methyl-2-phenyl-45-dihydrooxazol-4-yl)-2-phenylethyl]-diphenylphosphinite)(15-COD)iridium(I) tetrakis(35-bis(trifluoromethyl)phenylborate min. 97% (SS)-[COD]Ir[Ph2PThrePHOX]. MFCD06658140. Molecular Weight 1719.25. Molecular Formula Ir(C8H12)(C37H34NO2P)B[C6H3(CF3)2]4ae. Color/form orange pwdr. Strem 77-5020. http//www.strem.com/catalog/v/77-5020/