Learn More
Analytical Sales & Services SS Column Plug, 1/8"
Catalog No. 5000805019
Encompass
Stainless Steel Column Plug\n\n• 5/16-24 Thread, Coned Port\n• for 1/8" Tubing\n\nQty/Pkg: EACH
Catalog No. 50-008-05019 Supplier Analytical Sales & Services Supplier No. 1610
Stainless Steel Column Plug\n\n• 5/16-24 Thread, Coned Port\n• for 1/8" Tubing\n\nQty/Pkg: EACH