Learn More
Hach Company SS Conduc. Sen. boiler use 5.0
Catalog No. 5000207599 Shop All Hach Company Products
Encompass
Digital Stainless Steel Digital Contacting Conductivity Sensor for High Conductivity (k 5 0) - Boiler/CondensateNominal Cell Constant (K) 5 0 3/4 inch male NPT threaded 316 stainless steel body 6m (20 ft) Analog Cable Titanium Electrode Includes Digital Gateway and 1m (3 3 ft) Digital Extension Cable
Catalog No. 50-002-07599 Supplier Hach Company Supplier No. D3444D8
Digital Stainless Steel Digital Contacting Conductivity Sensor for High Conductivity (k 5 0) - Boiler/CondensateNominal Cell Constant (K) 5 0 3/4 inch male NPT threaded 316 stainless steel body 6m (20 ft) Analog Cable Titanium Electrode Includes Digital Gateway and 1m (3 3 ft) Digital Extension Cable