Learn More
Grainger SS Conductivity Sensor Tinned Leads
Catalog No. 502768166
Encompass_Preferred
The Grainger Approved SS Conductivity Sensor with Tinned Leads is a stainless steel conductivity sensor with a cell constant of 1.0 cm−1. It has a temperature range of 0 to 100°C, an immersion depth of 25 to 76 mm, and measures electrical conductivity in aqueous solutions. The sensor has a 316 stainless steel body, platinum electrodes, and 6 ft tinned copper lead wires. It can be used for water quality monitoring, agriculture, aquaculture, environmental studies, and other aqueous conductivity measurement applications.
Catalog No. 50-276-8166 Supplier Grainger Supplier No. 16Z061
The Grainger Approved SS Conductivity Sensor with Tinned Leads is a stainless steel conductivity sensor with a cell constant of 1.0 cm−1. It has a temperature range of 0 to 100°C, an immersion depth of 25 to 76 mm, and measures electrical conductivity in aqueous solutions. The sensor has a 316 stainless steel body, platinum electrodes, and 6 ft tinned copper lead wires. It can be used for water quality monitoring, agriculture, aquaculture, environmental studies, and other aqueous conductivity measurement applications.