Learn More
Hach Company SS Contact Conduct Sens-Low
Catalog No. 5000207997 Shop All Hach Company Products
Encompass
Stainless Steel Contacting Conductivity Sensor for Low Conductivity-k 0 5-Boiler/CondensateNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded 316 stainless steel body 6m (20 ft) Analog Cable Titanium Electrode
Catalog No. 50-002-07997 Supplier Hach Company Supplier No. 3444B8A
Stainless Steel Contacting Conductivity Sensor for Low Conductivity-k 0 5-Boiler/CondensateNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded 316 stainless steel body 6m (20 ft) Analog Cable Titanium Electrode