Learn More
Strem, An Ascensus Company SS cylinder 125ml 1cyl
SDP
Catalog No. 502034261
Encompass
CAS# . 1cyl. Stainless steel cylinder, 125ml, horizontal in line, with angled Bellows valve (150°C), DOT 4B, UN stamped. . Molecular Weight: . Molecular Formula: . Color/form: Strem# 95-0281. http://www.strem.com/catalog/v/95-0281/
Catalog No. 50-203-4261 Supplier Strem, An Ascensus Company Supplier No. 9502811CYL
CAS# . 1cyl. Stainless steel cylinder, 125ml, horizontal in line, with angled Bellows valve (150°C), DOT 4B, UN stamped. . Molecular Weight: . Molecular Formula: . Color/form: Strem# 95-0281. http://www.strem.com/catalog/v/95-0281/