Learn More
MSC Miller D-Bolt Anchorage Connector, Stainless Steel, 5/8 in. Bolt, 400 lb. Capacity, 1 ea.
Catalog No. 5000507532
Encompass
This stainless steel D-bolt anchorage connector is designed for secure, permanent installation in roofing applications. It provides a reliable anchor point for connecting fall protection devices, supporting a maximum working load of 400 lbs with a tensile strength of 5,000 lbs.
  • Constructed from durable stainless steel
  • Designed for permanent installation
  • Features a 5/8 in. bolt size
  • Compatible with 21/32 in. diameter holes
  • Meets OSHA 1910.66, OSHA 1926.502(d), ANSI A10.32, and ANSI Z359.1 requirements
  • Supports up to 5,000 lbs (22 kN) per worker
  • Requires user-supplied mounting hardware, including a 5/8 in. grade 8 bolt, lock washer, and nut
Catalog No. 50-005-07532 Supplier MSC Supplier No. 82609900
This stainless steel D-bolt anchorage connector is designed for secure, permanent installation in roofing applications. It provides a reliable anchor point for connecting fall protection devices, supporting a maximum working load of 400 lbs with a tensile strength of 5,000 lbs.
  • Constructed from durable stainless steel
  • Designed for permanent installation
  • Features a 5/8 in. bolt size
  • Compatible with 21/32 in. diameter holes
  • Meets OSHA 1910.66, OSHA 1926.502(d), ANSI A10.32, and ANSI Z359.1 requirements
  • Supports up to 5,000 lbs (22 kN) per worker
  • Requires user-supplied mounting hardware, including a 5/8 in. grade 8 bolt, lock washer, and nut