Learn More
Hach Company SS Dig Contct Condct Sens-High
Catalog No. 5000207154 Shop All Hach Company Products
Encompass
Stainless Steel Digital Contacting Conductivity Sensor for High Conductivity-k 5 0-Boiler/CondensateNominal Cell Constant (K) 5 0 3/4 inch male NPT threaded 316 stainless steel body 6m (20 ft) Analog Cable Titanium Electrode
Catalog No. 50-002-07154 Supplier Hach Company Supplier No. 3444D8A
Stainless Steel Digital Contacting Conductivity Sensor for High Conductivity-k 5 0-Boiler/CondensateNominal Cell Constant (K) 5 0 3/4 inch male NPT threaded 316 stainless steel body 6m (20 ft) Analog Cable Titanium Electrode