Learn More
Thermo Scientific Nano Bore Emitter Stainl. Steel 40mm OD 1/32
Catalog No. NC0379152 Shop All Thermo Scientific Products
Encompass
Electropolished, stainless steel LC/MS emitters, O.D. 150 um, I.D. 30 um. Each emitter is inserted into a microsleeve (I.D.180 um) to facilitate installation. Fits to capillary holder for 1/32. 4 pcs.
This item is not returnable. View return policy
Catalog No. NC0379152 Supplier Thermo Scientific Supplier No. ES542
Electropolished, stainless steel LC/MS emitters, O.D. 150 um, I.D. 30 um. Each emitter is inserted into a microsleeve (I.D.180 um) to facilitate installation. Fits to capillary holder for 1/32. 4 pcs.