Learn More
Ancare Corporation Stainless Steel Enrichment Cart for Storage and Washing of Enrichment Toys, Feeders, etc., 26” x 41” x 76” H
SDP
Catalog No. NC2745792
Encompass
Eight stainless steel wire baskets with lids (18” x 25” x 15”, #1 mesh).Two columns x four baskets high.Stainless steel welded solid top dust cover.Rear stops for baskets.Four 5” stainless steel casters with polyurethane wheels
Catalog No. NC2745792 Supplier Ancare Corporation Supplier No. ANRBWC2641SS8ST
Add to Cart
Add to Cart
Request a Quote
Eight stainless steel wire baskets with lids (18” x 25” x 15”, #1 mesh).Two columns x four baskets high.Stainless steel welded solid top dust cover.Rear stops for baskets.Four 5” stainless steel casters with polyurethane wheels