Learn More
MSC Precision Brand Feeler Gage, 300 Series Stainless Steel, 0.5 in. Wide, 12 in. Length, 12/Pk.
Catalog No. 5000515704
Encompass
This stainless steel feeler gage is designed for applications in manufacturing and maintenance environments requiring corrosion resistance and low magnetic properties to verify precise clearances and tight tolerances. Featuring a flat strip design, it is suitable for use in industries including aircraft, power, food, automotive, and marine. The material is a good conductor capable of withstanding high temperatures.
  • Constructed from 300 series stainless steel
  • Precision ground to close tolerances
  • Hardened and tempered to RC 48-62
  • Thickness clearly marked every 6 in. in metric and decimal
  • Meets ASTM A-666 chemistry standards and FED-GGG-G-17C
  • Dimensions: 0.5 in. wide by 12 in. long
Catalog No. 50-005-15704 Supplier MSC Supplier No. 99347551
This stainless steel feeler gage is designed for applications in manufacturing and maintenance environments requiring corrosion resistance and low magnetic properties to verify precise clearances and tight tolerances. Featuring a flat strip design, it is suitable for use in industries including aircraft, power, food, automotive, and marine. The material is a good conductor capable of withstanding high temperatures.
  • Constructed from 300 series stainless steel
  • Precision ground to close tolerances
  • Hardened and tempered to RC 48-62
  • Thickness clearly marked every 6 in. in metric and decimal
  • Meets ASTM A-666 chemistry standards and FED-GGG-G-17C
  • Dimensions: 0.5 in. wide by 12 in. long