Learn More
Waters Corp Ferrule, Annealed, Stainless Steel, 1/16 in
Catalog No. NC3404897 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a standard stainless steel ferrule designed for use as a fitting in HPLC systems.
  • Designed as a fitting
  • Suitable for HPLC systems
  • Constructed from stainless steel
  • Features an internal diameter of 1.5875 mm
  • Can be replaced as required
Catalog No. NC3404897 Supplier Waters Corp Supplier No. P645512
This is a standard stainless steel ferrule designed for use as a fitting in HPLC systems.
  • Designed as a fitting
  • Suitable for HPLC systems
  • Constructed from stainless steel
  • Features an internal diameter of 1.5875 mm
  • Can be replaced as required